Learn More
Medchemexpress LLC Sericin | 250 MG
SDP
Catalog No. 5000426744
Encompass_Preferred
Sericin, isolated from Bombyx mori silk cocoons, is a spherical protein known for its diverse biological activities. It acts as a cognitive enhancer, pain reliever, and can serve as a cryoprotectant agent replacing fetal bovine serum or DMSO. Sericin helps reduce oxidative stress and reactive oxygen species, and facilitates wound repair through collagen production.
  • Exhibits antioxidant properties
  • Offers anti-diabetic benefits
  • Shows anti-hyperlipidemic effects
  • Provides anti-inflammatory action
  • Acts as a skin moisturizer
  • Promotes wound healing
  • Possesses antibacterial and antimicrobial qualities
  • Demonstrates anti-tumor protective properties
Catalog No. 50-004-26744 Supplier Medchemexpress LLC Supplier No. 250MGHYNP192
Sericin, isolated from Bombyx mori silk cocoons, is a spherical protein known for its diverse biological activities. It acts as a cognitive enhancer, pain reliever, and can serve as a cryoprotectant agent replacing fetal bovine serum or DMSO. Sericin helps reduce oxidative stress and reactive oxygen species, and facilitates wound repair through collagen production.
  • Exhibits antioxidant properties
  • Offers anti-diabetic benefits
  • Shows anti-hyperlipidemic effects
  • Provides anti-inflammatory action
  • Acts as a skin moisturizer
  • Promotes wound healing
  • Possesses antibacterial and antimicrobial qualities
  • Demonstrates anti-tumor protective properties