Learn More
TARGETMOL CHEMICALS INC SERIDOPIDINE 5MG
SDP
Catalog No. 502580253
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Seridopidine (ACR343) is a modulator of dopaminergic activity that is used as an oral therapy for schizophrenia Parkinson's disease and Tourette's syndrome. purity: 99%
Catalog No. 50-258-0253 Supplier TARGETMOL CHEMICALS INC Supplier No. T346165MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Seridopidine (ACR343) is a modulator of dopaminergic activity that is used as an oral therapy for schizophrenia Parkinson's disease and Tourette's syndrome. purity: 99%