Learn More
Epredia™ Series 110 Laser Slotted with Front Tab Cassettes with Lids
Catalog No. 1100410LMI Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11-004-10LMI Pink
11-011-10LMI Ochre
11-012-10LMI Red
11-010-10LMI Beige
11-003-10LMI Blue
11-001-10LMI White
11-005-10LMI Green
11-006-10LMI Orange
11-002-10LMI Yellow
11-007-10LMI Gray
11-008-10LMI Aqua
11-009-10LMI Lilac
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 11-004-10LMI Supplier Epredia™ Supplier No. 1100410LMI
Add to Cart
Edge
Add to Cart
Manufactured specifically for laser printing, the Epredia™ Series 110 Slotted Cassettes come with separate lids with a tab in the front for easy opening and closing.
  • Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
  • Contain a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Each style comes in different colors with color-matched lids
  • Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Comes in taped stacks of 40 cassettes; 25 stacks per case (for a total of 1000 cassettes with separate lids)
  • The Epredia™ Laser Cassette is not compatible with thermal tape style printers
  • Tape strips on the stacks is made from 95% recycled polypropylene and is entirely recyclable
Specifications
Pore Style Slots
Quantity 40 Cassettes/Taped Stack, 25 Stacks/Cs.
Closure Included Yes
Closure Type Separate Lid with Tab
Color Pink
Product Type Cassette