Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Series 210 Laser Biopsy Cassettes with Front Tab Lids

Catalog No. 2101110LM Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-011-10LM Ochre
21-001-10LM White
21-002-10LM Yellow
21-003-10LM Blue
21-004-10LM Pink
21-005-10LM Green
21-006-10LM Orange
21-007-10LM Gray
21-008-10LM Aqua
21-009-10LM Lilac
21-010-10LM Beige
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 21-011-10LM Supplier Epredia™ Supplier No. 2101110LM

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured specifically for laser printing, the Series 210 Biopsy Cassettes come with separate lids with a tab in the front for easy opening and closing.

  • Epredia™ Laser Cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ Laser Printers
  • Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Comes in multiple different colors with color-matched lids
  • Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Comes in taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes with separate lids
  • Not compatible with thermal tape style printers

Specifications

Pore Style Small Squares
Quantity 40 Cassettes/Taped Stack, 25 Stacks/Cs.
Closure Included Yes
Closure Type Separate Lid with Tab
Color Ochre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.