Learn More
Hach Company Series 5000 Silica Analyser Reagent Set, with Silica Standard
Catalog No. NC1900903 Shop All Hach Company Products
Encompass
Series 5000 Silica Analyser Reagent Set, with Silica Standard. Contains Molybdate 3 Reagent (2.9 L), Citric Acid/Surfactant Reagent (2.9 L), Amino Acid F Reagent (2.9 L), and Silica Standard Solution, 500.00 µg/L SiO2 (2.9 L).
Catalog No. NC1900903 Supplier Hach Company Supplier No. 4562700
Series 5000 Silica Analyser Reagent Set, with Silica Standard. Contains Molybdate 3 Reagent (2.9 L), Citric Acid/Surfactant Reagent (2.9 L), Amino Acid F Reagent (2.9 L), and Silica Standard Solution, 500.00 µg/L SiO2 (2.9 L).