Learn More
Description
- Epredia™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Each style comes in multiple different colors
- Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Series 815 come in taped stacks of 40 cassettes with lids attached; 45 stacks per case for a total of 1800 cassettes
- The Epredia™ Laser Cassette is not compatible with thermal tape style printers
- Tape strips on the stacks is made from 95% recycled polypropylene and is entirely recyclable
Specifications
Specifications
| Pore Style | Slots |
| Quantity | 40 Cassettes/Taped Stack, 45 Stacks/Cs. |
| Closure Included | Yes |
| Closure Type | Attached Lid |
| Color | Green |
| Product Type | Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.