Learn More
BINDER™ Series FP Avantgarde.Line Drying and Heating Chambers, Gas-tight Option
SDP
Catalog No. 08101229 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Series FP 056 Avantgarde
BINDER Series FP 115 Avantgarde
BINDER Series FP 260 Avantgarde
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
08-101-229 BINDER Series FP 115 Avantgarde
08-101-230 BINDER Series FP 260 Avantgarde
08-101-228 BINDER Series FP 056 Avantgarde
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 08-101-229 Supplier BINDER™ Supplier No. 80122217
Add to Cart
Add to Cart
This item is not returnable. View return policy
The gas tight option model for Series FP Avantgarde.Line Drying and Heating Chambers provides additional measures for the greatest possible impermeability to gas.
  • Accessory for Series FP Avantgarde.Line Drying and Heating Chambers
Specifications
Description Not in conjunction with access port or interior lighting option.
For Use With (Equipment) BINDER Series FP 115 Avantgarde
Type Drying and Heating Chamber
Model Gas-Tight Option
Voltage 120/230 V