Learn More
Perkin Elmer US LLC Series II Ion Lens Removal Tool for ELAN 6x00/9000/DRCs (Post-1998/Pre-2005)
Catalog No. 502113111
Encompass_Preferred
Flexible shaft driver with removable 2 mm Hex Ball Bit for use with ELAN 6x00, 9000, DRC-e and DRC II ICP-MS series instruments manufactured between 1998 and 2005.
Catalog No. 50-211-3111 Supplier Perkin Elmer US LLC Supplier No. W1010620
Flexible shaft driver with removable 2 mm Hex Ball Bit for use with ELAN 6x00, 9000, DRC-e and DRC II ICP-MS series instruments manufactured between 1998 and 2005.