Learn More
Perkin Elmer US LLC Series II Ion Lens for ELAN 6x00/9000/DRCs (Post-1998)
Catalog No. 509049520
Encompass_Preferred
PerkinElmer offers the industry’s only single-ion lens with the exclusive SwiftMount™ II cassette lens mount. No tools are required as the design incorporates a clean capture-and-release concept.
Catalog No. 50-904-9520 Supplier Perkin Elmer US LLC Supplier No. WE018034
PerkinElmer offers the industry’s only single-ion lens with the exclusive SwiftMount™ II cassette lens mount. No tools are required as the design incorporates a clean capture-and-release concept.