Learn More
Mark 10 Corporation Series TT01 Cap Torque Testers (50 lbFin / 570 Ncm Capacity
SDP
Catalog No. NC4019238
Encompass
Series TT01 Cap Torque Testers are designed to measure application and removal torque of bottle caps. Adjustable posts effectively grip a broad range of container shapes and sizes, while optional flat and adjustable jaws are available for unique profiles. A rugged aluminum design, and simple, intuitive operation allow for use in laboratory and production environments.
Catalog No. NC4019238 Supplier Mark 10 Corporation Supplier No. MTT0150
Series TT01 Cap Torque Testers are designed to measure application and removal torque of bottle caps. Adjustable posts effectively grip a broad range of container shapes and sizes, while optional flat and adjustable jaws are available for unique profiles. A rugged aluminum design, and simple, intuitive operation allow for use in laboratory and production environments.