Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Serine Hydrolase Inhibitor-20

Catalog No. ALD005445MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD005445MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. ALD005445MG Supplier Sigma Aldrich Supplier No. ALD005445MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Serine hydrolase inhibitors are small molecule fragments that react in vitro with the serine hydrolase class of proteins. They are amenable to further modification to create novel inhibitors of this class of protein.

Specifications

CAS 2074732-95-7
Molecular Formula C13H20N2O
Linear Formula C13H20N2O
Quantity 50 mg
Synonym Yu MPAA mLigand
Molecular Weight (g/mol) 220.31 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.