Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Serine Inhibitor Cocktail I, for Broad Range Serine Proteases, 1 Vial
SDP

Catalog No. 50488964
Encompass_Preferred

Mixture of four protease inhibitors for inhibiting serine proteases. Supplied in 1ml DMSO.

Contains:

AEBSF
Aprotinin
Elastatinal
GGACK

Specificity:
Chymotrypsin
Kallikrein
Plasmin Thrombin
Trypsin
Elastase
Urokinase
Factor Xa

Catalog No. 50-488-964 Supplier Research Products International Corp Supplier No. P516001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mixture of four protease inhibitors for inhibiting serine proteases. Supplied in 1ml DMSO.

Contains:

AEBSF
Aprotinin
Elastatinal
GGACK

Specificity:
Chymotrypsin
Kallikrein
Plasmin Thrombin
Trypsin
Elastase
Urokinase
Factor Xa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.