Learn More
Abcam Serine Protease Assay Kit (FAM-Spacer-Leu-CMK), 25 Tests
Catalog No. 502683252
Encompass_Preferred
These Serine Protease Assay Kits enable researchers to detect intracellular chymotrypsin-like serine protease activity in vitro without lysing the cell.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-3252 Supplier Abcam Supplier No. AB27077825TEST
These Serine Protease Assay Kits enable researchers to detect intracellular chymotrypsin-like serine protease activity in vitro without lysing the cell.
The product is subject to the following: Abcam Restricted Use Statement