Learn More
Abcam Serine Protease Assay Kit (FAM-Spacer-Leu-CMK), 100 Tests
Catalog No. 502683253
Encompass_Preferred
These Serine Protease Assay Kits enable researchers to detect intracellular chymotrypsin-like serine protease activity in vitro without lysing the cell.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-3253 Supplier Abcam Supplier No. AB270778100TEST
These Serine Protease Assay Kits enable researchers to detect intracellular chymotrypsin-like serine protease activity in vitro without lysing the cell.
The product is subject to the following: Abcam Restricted Use Statement