Learn More
Abcam Serine Protease Assay Kit (SR101-Leu-CMK), 100 Tests
Catalog No. 502671982
Encompass_Preferred
These Serine Protease Assay Kits enable researchers to detect intracellular chymotrypsin-like serine protease activity in vitro without lysing the cell.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1982 Supplier Abcam Supplier No. AB270776100TEST
These Serine Protease Assay Kits enable researchers to detect intracellular chymotrypsin-like serine protease activity in vitro without lysing the cell.
The product is subject to the following: Abcam Restricted Use Statement