Learn More
AG Scientific Inc Serine Protease Inhib I
Catalog No. 502522610
Encompass_Preferred
"Serine Protease Inhibitor Cocktail I is a mixture of four protease inhibitors which has been optimized for the inhibition of a broad range of serine proteases. Reconstitute each vial with 100 mL water to obtain a 1X concentrated stock solution. 1X stock solution contains: 500uM AEBSF HCl 420 nM Aprotinin 20uM Elastatinal 1uM GGACK​​​​​"
Catalog No. 50-252-2610 Supplier AG Scientific Inc Supplier No. P1516VIAL
Add to Cart
Add to Cart
"Serine Protease Inhibitor Cocktail I is a mixture of four protease inhibitors which has been optimized for the inhibition of a broad range of serine proteases. Reconstitute each vial with 100 mL water to obtain a 1X concentrated stock solution. 1X stock solution contains: 500uM AEBSF HCl 420 nM Aprotinin 20uM Elastatinal 1uM GGACK​​​​​"