Learn More
AdipoGen Seriniquinone
SDP
Supplier:  AdipoGen BVT0478M001
Encompass_Preferred
Chemical. CAS 22200-69-7. Formula C20H8O4S. MW 344.3. Synthetic. Originally isolated from marine bacterium Serinicoccus sp. strain CNJ927. Seriniquinone is a potent cytotoxic and melanoma-selective anticancer agent. Upon cellular treatment and based on the compounds fluorescent nature it was found localized in the endoplasmic reticulum. Seriniquinone inhibits cell proliferation marked by autophagocytosis and induces cell death due to caspase-9 dependent apoptosis. Mode of action studies suggest that it specifically targets a skin protective antimicrobial peptide dermcidin DCD, whose role as a cancer-protective protein has been recently recognized.
Catalog No. 50-256-1881