Learn More
Bellco Glass, Inc. Serological Cotton Plug Top Pipette,10x.1mL-Bulk
SDP
Catalog No. 501944574
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class A Borosilicate glass. All Bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Constructed top for cotton plugging (supplied without cotton). Calibrated to tip -tempered tip; to deliver; blow out. MADE IN THE USACase Qty: 250TOLERANCE +-(ML):0.06OAL (MM):350COLOR CODE: Orange Graduation: 0.1
Catalog No. 50-194-4574 Supplier Bellco Glass, Inc. Supplier No. 120010010
Bellco manufactures all precision pipettes from type 1 class A Borosilicate glass. All Bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Constructed top for cotton plugging (supplied without cotton). Calibrated to tip -tempered tip; to deliver; blow out. MADE IN THE USACase Qty: 250TOLERANCE +-(ML):0.06OAL (MM):350COLOR CODE: Orange Graduation: 0.1