Learn More
Bellco Glass, Inc. Serological Cotton Plug Top Pipette,5x.1mL
SDP
Catalog No. 501944590
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Constricted top for cotton plugging (supplied with out cotton). MADE IN THE USA Case Qty: 18 TOLERANCE +-(ML):0.04OAL (MM):350COLOR CODE: Blue Graduation: 0.1
Catalog No. 50-194-4590 Supplier Bellco Glass, Inc. Supplier No. 121005010
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Constricted top for cotton plugging (supplied with out cotton). MADE IN THE USA Case Qty: 18 TOLERANCE +-(ML):0.04OAL (MM):350COLOR CODE: Blue Graduation: 0.1