Learn More
Bellco Glass, Inc. Serological Cotton Plug Top Pipette Mix,1/5/10-Bulk
SDP
Catalog No. 501944571
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class A Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Constructed top for cotton plugging (supplied without cotton). Calibrated to tip -tempered tip; to deliver; blow out. MADE IN THE USA75 each 1ML, 5ML and 100 each 10MLColor code:1ml – yellow5ml – blue10ml – orangeCase Qty: 250OAL (MM):350 Graduations: 1mL - .01 5mL - 0.1 10mL - 0.1
Catalog No. 50-194-4571 Supplier Bellco Glass, Inc. Supplier No. 120001510
Bellco manufactures all precision pipettes from type 1 class A Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Constructed top for cotton plugging (supplied without cotton). Calibrated to tip -tempered tip; to deliver; blow out. MADE IN THE USA75 each 1ML, 5ML and 100 each 10MLColor code:1ml – yellow5ml – blue10ml – orangeCase Qty: 250OAL (MM):350 Graduations: 1mL - .01 5mL - 0.1 10mL - 0.1