Learn More
Bellco Glass, Inc. Serological Pipet Large Orfice,50 x .2mL Cotton Plug Top
SDP
Catalog No. 501944642
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. 3mm orifice for handling tissue homogenizer. Borosilicate Glass. MADE IN THE USA Case qty-12 TOLERANCE +-(ML):0.5OAL (MM):470COLOR CODE: Red Graduation: 0.2
Catalog No. 50-194-4642 Supplier Bellco Glass, Inc. Supplier No. 123250020
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. 3mm orifice for handling tissue homogenizer. Borosilicate Glass. MADE IN THE USA Case qty-12 TOLERANCE +-(ML):0.5OAL (MM):470COLOR CODE: Red Graduation: 0.2