Learn More
Foxx Life Sciences Borosil® Reusable Class B Serological Pipettes (Pipets) with Permanent Amber Graduations, 2mL (0.02 Intervals), 20/CS
Catalog No. 501720421
Encompass_Preferred
Borosil® Reusable Class B Serological Pipettes are graduated using a permanent amber treatment that will not wear off even after reuse. Manufactured with consistent wall thickness. Reusable. Uniform inner bore diameter for accuracy between different readings. Beveled tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil® Serological Pipettes are graduated for delivery from the zero mark to the tip, without "blowing out" the droplet remaining in the tip. Borosil® Class B Serological Pipettes comply with EN ISO 835. Available from 1mL to 25mL. 1mL,2mL and 5mL size available in two different graduation intervals. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices.
Catalog No. 50-172-0421 Supplier Foxx Life Sciences Supplier No. 7080P02D
Borosil® Reusable Class B Serological Pipettes are graduated using a permanent amber treatment that will not wear off even after reuse. Manufactured with consistent wall thickness. Reusable. Uniform inner bore diameter for accuracy between different readings. Beveled tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil® Serological Pipettes are graduated for delivery from the zero mark to the tip, without "blowing out" the droplet remaining in the tip. Borosil® Class B Serological Pipettes comply with EN ISO 835. Available from 1mL to 25mL. 1mL,2mL and 5mL size available in two different graduation intervals. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices.