Learn More
Chemglass Life Sciences Serological Pipette, 1mL, Color Coded Yellow, Individually Wrapped, Sterile
SDP
Catalog No. 501897697
Encompass_Preferred
Serological pipettes are made of super clear clarity polystyrene (PS). Serological pipettes are designed to be used in both tissue culture and bacterial culture experiments, and other liquid handling applications.Edged color-coded band for identificationInclude negative graduations for extra volumeNon-pyrogenic, non-cytotoxic, non-hemolytic, DNase- and RNase-freePolyolefin fiber filter to reduce contaminationMade from high clarity polystyreneSterilized by E-beam
Catalog No. 50-189-7697 Supplier Chemglass Life Sciences Supplier No. CGN3481001
Serological pipettes are made of super clear clarity polystyrene (PS). Serological pipettes are designed to be used in both tissue culture and bacterial culture experiments, and other liquid handling applications.Edged color-coded band for identificationInclude negative graduations for extra volumeNon-pyrogenic, non-cytotoxic, non-hemolytic, DNase- and RNase-freePolyolefin fiber filter to reduce contaminationMade from high clarity polystyreneSterilized by E-beam