Learn More
Bellco Glass, Inc. Serological Plain Top Pipette,2x.1mL-Bulk
SDP
Supplier:  Bellco Glass, Inc. 120202010
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Plain top promotes rapid even flow with a calibrated to tip -tempered tip; todeliver; blow out. MADE IN THE USA Case Qty: 250 TOLERANCE +-(ML):0.02OAL (MM):350COLOR CODE: Green Graduation: 0.1
Catalog No. 50-194-4578