Learn More
Bellco Glass, Inc. Serological Plain Top Pipette, 2x.1mL
SDP
Supplier:  Bellco Glass, Inc. 121202010
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Plain top promotes rapid & even flow. MADE IN THE USA Case Qty: 18 TOLERANCE +-(ML):0.02OAL (MM):350COLOR CODE: Green Graduation: 0.1
Catalog No. 50-194-4595