Learn More
Bellco Glass, Inc. Serological Plain Top Pipette, 5x.1mL
SDP
Catalog No. 501944596
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Plain top promotes rapid & even flow. MADE IN THE USA Case Qty: 18 TOLERANCE +-(ML):0.04OAL (MM):350COLOR CODE: Blue Graduation: 0.1
Catalog No. 50-194-4596 Supplier Bellco Glass, Inc. Supplier No. 121205010
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Plain top promotes rapid & even flow. MADE IN THE USA Case Qty: 18 TOLERANCE +-(ML):0.04OAL (MM):350COLOR CODE: Blue Graduation: 0.1