Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix SeroDetect Anti-HBs Panel
SDP

Catalog No. 22156707 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-707 6 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product capable of transmitting infectious agents
Catalog No. 22-156-707 Supplier ZeptoMetrix Supplier No. KZMC008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product capable of transmitting infectious agents

For monitoring assay performance across wide range of reactivity levels

  • Composed of six members
  • One vial (1.5mL) SeroDetect Anti-HBs, non-reactive for antibodies to Hepatitis B Surface Antigen and five vials (1.5mL) containing antibodies to Hepatitis B Surface Antigen
  • Prepared from proteins derived from human sources, and non human components (antimicrobial agents and stabilizers)
  • Processed to eliminate contaminants
  • Needs evaluation of members as per instructions

Not substitute for mandatory positive and negative control reagents provided with licensed test kits

For training, lot-to-lot comparison of reagent test kits and to evaluate and compare intra laboratory and inter laboratory performance of Anti-HBs test systems

Specifications

Content And Storage 2°C to 8°C
Product Type SeroDetect Anti-HBs Panel
Quantity 6 x 1.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.