Learn More
Description
Includes
- Reagents
- Two vials SeroDetect Dengue Fever Negative (0.5mL each)
- Eight vials SeroDetect Dengue Fever Positive (0.5mL each)
Designed for use with in vitro assay procedures
Materials prepared from human source serum and other non-human components
Serum is heat inactivated to reduce infectious risk
For research use only
For training
Comparision of reagent test kits
Evaluation and comparison of intra and inter laboratory Dengue Fever test system performance
Specifications
Specifications
| Biosafety Level | 2 |
| Content And Storage | 2°C to 8°C |
| Product Type | Microbiology Media |
| For Use With (Application) | For Research Use only |
| Quantity | 0.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
