Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix SeroDetect HIV-1/ HIV-2 Ag/Ab Combo Verification Panel
SDP

Catalog No. 22156108 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
5 x 1.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-108 5 x 1.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-108 Supplier ZeptoMetrix Supplier No. KZMC030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with assays in the determination of the presence of antigens and antibodies to HIV-1 and HIV-2

  • Composed of five members representing a variety of reactivities
  • Prepared from citrated human plasma
  • Can be used for training, lot-to-lot comparison of reagent test kits as well as evaluation and comparison of intra- and inter-laboratory performance of HIV-1 and HIV-2 Ag/Ab test systems

Specifications

Certifications/Compliance FDA
Content And Storage 2°C to 8°C
Product Type HIV-1/ HIV-2 Ag/Ab Combo Verification Panel
For Use With (Application) Determine presence of antigens and antibodies to HIV-1 and HIV-2
Quantity 5 x 1.25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.