Learn More
Description
Includes
Reagents
- One vial SeroDetect HIV-Antibody, Negative (1.5mL)
- Four vials SeroDetect HIV-Antibody, Positive (1.5mL each)
- Specimens contain proteins derived from human sources, antimicrobial agents and stabilizers
- Five specimens, 1.5mL each
Used for training, lot to lot comparison of reagent test kits, evaluate and compare intralaboratory and inter laboratory performance of HIV-Antibody test systems.
Specifications
Specifications
| Biosafety Level | 2 |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | SeroDetect HIV Antibody Panel |
| For Use With (Application) | In vitro assay procedures for the determination of Antibodies to HIV Types 1 and 2. |
| Quantity | 5 x 1.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
