Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fujirebio Diagnostics Inc SERODIA TP•PA Kit, Passive Particle Agglutination, 100 tests, 2 hours incubation, Human serum or plasma, High sensitivity, High specificity

Supplier:  Fujirebio Diagnostics Inc 234952

Encompass

The SERODIA TP•PA Kit is a diagnostic test designed for the detection of *Treponema pallidum* antibodies in human serum or plasma. Utilizing a qualitative and semi-quantitative passive particle agglutination method, this kit aids in the diagnosis of syphilis by allowing for visual interpretation of results after a two-hour incubation. The assay is specifically engineered to minimize non-specific agglutination and does not require any specialized equipment, ensuring ease of use in various settings.

  • Test type: Passive particle agglutination
  • Detection target: *Treponema pallidum* antibodies
  • Specimen type: Human serum or plasma
  • Incubation time: 2 hours
  • Reading method: Visual interpretation
  • Equipment required: No special equipment
  • Specificity: High
  • Sensitivity: High
  • Application: Aid in diagnosis of syphilis
  • Preservative: Sodium azide
  • Kit size: 100 tests

Catalog No. NC2940996


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.