Learn More
Vee Gee Vee Gee Serological Pipet, 10 mL, 0.1 graduation interval
Catalog No. NC1937450
Encompass_Preferred
The Serological Pipet delivers and blows out. It meets ASTM E-1044 Style 1, Class A Specifications.: 10.0 mL Tolerance: 0.03 mL Graduation Interval: 0.1 mL Approx. O.D. x Height: 10 x 370 mm Color Code: Orange
Catalog No. NC1937450 Supplier Vee Gee Supplier No. 2011A10
The Serological Pipet delivers and blows out. It meets ASTM E-1044 Style 1, Class A Specifications.: 10.0 mL Tolerance: 0.03 mL Graduation Interval: 0.1 mL Approx. O.D. x Height: 10 x 370 mm Color Code: Orange