Learn More
Globe Scientific Serological Pipet Storage Rack
Catalog No. 21402960
Change view
Click to view available options
Color:
Blue
Red
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-960 Blue
21-402-961 Red
21-402-962 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-402-960 Supplier Globe Scientific Supplier No. 1685B
Only null left
Add to Cart
Add to Cart
Four angled compartments protect and organize glass or plastic serological pipets
  • Durable ABS construction for scratch resistance and easy cleaning
  • Strong magnets on both sides for secure mounting to most metal surfaces
  • Rack can stand on flat surfaces without mounting
  • Two included accessory attachments keep pipet fillers and bulbs within reach
  • Side windows allow for easy viewing of pipet size and inventory
Specifications
Autoclavable Not Autoclavable
Color Blue
Holds Serological Pipets
Material ABS with Mounting Magnets
Positions 4
Shape Rectangular
Stackable No
Sterility Non-sterile
Product Type Side-mounted Pipet Holder
Shelves 4 Tiers
Height (Metric) 395 mm
Array 1 x 4
Width (Metric) 85 mm