Learn More
Mascon Medical Serological Pipettes, Sterile, Polystyrene, 10ml, 1EA/Bag, 200EA/Case
SDP
Catalog No. NC2180292
Encompass_Preferred
Mascon Medical™ Serological Pipettes are manufactured with a durable welded 3-piece construction and supplied with a cotton plug to help to prevent overfill. These pipettes are made from virgin polystyrene and are ideal for accurate liquid transfer or mixing. They are also color coded with black ink graduations for ease of use (accurate 2% at full volume).Specifications: Volume: 10ml Qty. / Case: 200 Material: Virgin Polystyrene Plug Material: Cotton Color: Purple Sterile: Yes Packaging: Individually Packed Packaging Format: 1EA/Bag, 200EA/CaseFeatures: Accurate 2% at full volume Black ink graduations Color coded Cotton plug
This item is not returnable. View return policy
Catalog No. NC2180292 Supplier Mascon Medical Supplier No. FSSLPP10
Mascon Medical™ Serological Pipettes are manufactured with a durable welded 3-piece construction and supplied with a cotton plug to help to prevent overfill. These pipettes are made from virgin polystyrene and are ideal for accurate liquid transfer or mixing. They are also color coded with black ink graduations for ease of use (accurate 2% at full volume).Specifications: Volume: 10ml Qty. / Case: 200 Material: Virgin Polystyrene Plug Material: Cotton Color: Purple Sterile: Yes Packaging: Individually Packed Packaging Format: 1EA/Bag, 200EA/CaseFeatures: Accurate 2% at full volume Black ink graduations Color coded Cotton plug