Learn More
Foxx Life Sciences Borosil Reusable Class A Serological Pipettes (Pipets) with Permanent Amber Graduations and Individual Serial Numbers, Batch Certificate, 10mL, 10/CS
Catalog No. 501542885
Encompass_Preferred
Borosil Reusable Class A Serological Pipettes are graduated using a permanent amber treatment that will not wear off even after reuse. Each pipette is individually calibrated by automatic, laser-controlled calibration equipment. Manufactured with consistent wall thickness. Reusable. Uniform inner bore diameter for accuracy between different readings. Beveled tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil Serological Pipettes are graduated for delivery from the zero mark to the tip, without "blowing out" the droplet remaining in the tip. Borosil Class A Serological Pipettes comply with ASTM 1293-02. Avail. from 1mL to 25mL. 1mL size Avail. in two different graduation intervals (0.01mL & 0.1mL). Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices.
Catalog No. 50-154-2885 Supplier Foxx Life Sciences Supplier No. 7081P06A
Borosil Reusable Class A Serological Pipettes are graduated using a permanent amber treatment that will not wear off even after reuse. Each pipette is individually calibrated by automatic, laser-controlled calibration equipment. Manufactured with consistent wall thickness. Reusable. Uniform inner bore diameter for accuracy between different readings. Beveled tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil Serological Pipettes are graduated for delivery from the zero mark to the tip, without "blowing out" the droplet remaining in the tip. Borosil Class A Serological Pipettes comply with ASTM 1293-02. Avail. from 1mL to 25mL. 1mL size Avail. in two different graduation intervals (0.01mL & 0.1mL). Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices.