Learn More
Foxx Life Sciences Borosil® Reusable Class A Serological Pipettes (Pipets) with Permanent Amber Graduations and Individual Serial Numbers, Individual Certificate, 1mL (0.1mL Intervals), 10/CS
Catalog No. 501720318
Encompass_Preferred
Borosil® Reusable Class A Serological Pipettes are graduated using a permanent amber treatment that will not wear off even after reuse. Each pipette is individually calibrated by automatic, laser-controlled calibration equipment. Manuf.w/ consistent wall thickness. Reusable. Uniform inner bore diameter for accuracy between different readings. Beveled tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil® Serological Pipettes are graduated for delivery from the zero mark to the tip, without "blowing out" the droplet remaining in the tip. Borosil® Class A Serological Pipettes comply w/ ASTM 1293-02. Available from 1mL to 25mL. 1mL size available in two different graduation intervals (0.01mL & 0.1mL). Manuf.from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices.
Catalog No. 50-172-0318 Supplier Foxx Life Sciences Supplier No. 7081P11D
Borosil® Reusable Class A Serological Pipettes are graduated using a permanent amber treatment that will not wear off even after reuse. Each pipette is individually calibrated by automatic, laser-controlled calibration equipment. Manuf.w/ consistent wall thickness. Reusable. Uniform inner bore diameter for accuracy between different readings. Beveled tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil® Serological Pipettes are graduated for delivery from the zero mark to the tip, without "blowing out" the droplet remaining in the tip. Borosil® Class A Serological Pipettes comply w/ ASTM 1293-02. Available from 1mL to 25mL. 1mL size available in two different graduation intervals (0.01mL & 0.1mL). Manuf.from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices.