Learn More
LUNA NANOTECH INC Seropal Serological Pipette Controller
Catalog No. NC3667884
Encompass_Preferred
SeroPAL™ is an ergonomically designed electronic serological pipette controller capable of pipetting volumes of liquids between 0.1 mL and 100 mL. 
Features:
  • Handles volumes between 0.1 mL and 100 mL
  • Works with plastic and glass serological pipettes
  • Includes a wall or bench mountable holder for the controller
  • Motor speed controller allows selection of the pipetting speed
  • Mode selection switch allows switching between gravity based and motor driven liquid dispensing
  • Direct air inlet/exhaust protects against corrosion of internal parts
  • Membrane filter prevents contamination of internal parts of the controller
  • Battery recharging in just 2 hours providing 10 hours of continuous operation
  • Specifically designed electronic system prevents lithium ion battery overcharging
  • Highly ergonomic design
  • Lithium battery cover i
Catalog No. NC3667884 Supplier LUNA NANOTECH INC Supplier No. SEP100S
SeroPAL™ is an ergonomically designed electronic serological pipette controller capable of pipetting volumes of liquids between 0.1 mL and 100 mL. 
Features:
  • Handles volumes between 0.1 mL and 100 mL
  • Works with plastic and glass serological pipettes
  • Includes a wall or bench mountable holder for the controller
  • Motor speed controller allows selection of the pipetting speed
  • Mode selection switch allows switching between gravity based and motor driven liquid dispensing
  • Direct air inlet/exhaust protects against corrosion of internal parts
  • Membrane filter prevents contamination of internal parts of the controller
  • Battery recharging in just 2 hours providing 10 hours of continuous operation
  • Specifically designed electronic system prevents lithium ion battery overcharging
  • Highly ergonomic design
  • Lithium battery cover i