Learn More
Enzo Life Sciences SCREEN-WELL Serotonergic ligand library (500 µl/well)
SDP
Catalog No. 502010193
Encompass_Preferred
The Serotonergic Ligands Plate contains 79 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Serotonergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 680 ligands. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0193 Supplier Enzo Life Sciences Supplier No. BML28130500
Add to Cart
Add to Cart
Request a Quote
The Serotonergic Ligands Plate contains 79 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Serotonergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 680 ligands. Long Term Storage: -80°C. Applications: HTS .