Learn More
Selleck Chemical LLC Serotonin (5-HT) HCl S4244-1g
SDP
Catalog No. 503109951
Encompass_Preferred
Serotonin (5-HT) HCl is a monoamine neurotransmitter and Endogenous 5-HT receptor agonist
Catalog No. 50-310-9951 Supplier Selleck Chemical LLC Supplier No. S42441G
Serotonin (5-HT) HCl is a monoamine neurotransmitter and Endogenous 5-HT receptor agonist