Learn More
Apexbio Technology LLC SEROTONIN-1MG
SDP
Catalog No. 5000570507
Encompass_Preferred
Serotonin (CAS No 50-67-9) is a neurotransmitter primarily involved in regulating mood cognition and various physiological processes Originating from the amino acid tryptophan serotonin s mechanism of action involves binding to its receptors influencing several intracellular pathways including cyclic AMP and diacylglycerol In biomedical research its in vitro activity is commonly observed in the nanomolar to low micromolar range Serotonin is widely used in studies focusing on neurological and psychiatric conditions as well as in drug screening assays Typical applications include the use of cell models and animal studies to explore its effects on brain function and behavior Experimental concentrations typically depend on the specific research design allowing flexibility in adapting the dosages to various models and hypotheses
Catalog No. 50-005-70507 Supplier Apexbio Technology LLC Supplier No. BA49891
Serotonin (CAS No 50-67-9) is a neurotransmitter primarily involved in regulating mood cognition and various physiological processes Originating from the amino acid tryptophan serotonin s mechanism of action involves binding to its receptors influencing several intracellular pathways including cyclic AMP and diacylglycerol In biomedical research its in vitro activity is commonly observed in the nanomolar to low micromolar range Serotonin is widely used in studies focusing on neurological and psychiatric conditions as well as in drug screening assays Typical applications include the use of cell models and animal studies to explore its effects on brain function and behavior Experimental concentrations typically depend on the specific research design allowing flexibility in adapting the dosages to various models and hypotheses