Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Serotonin (5-hydroxytryptamine) | 50-67-9 | MFCD00055054 | 99.3% | 176.21 g/mol | C10H12N2O | 10 MG
SDP

Catalog No. 5000298897
Encompass_Preferred

Serotonin (5-hydroxytryptamine) is an endogenous monoamine neurotransmitter and 5-HT receptor agonist commonly used as a biochemical reagent and analytical standard. It is supplied as a high-purity solid suitable for receptor pharmacology, analytical method development, and biochemical assays.

  • High purity (99.26%) suitable for analytical work.
  • Available as a solid in 10 mg quantities.
  • CAS number 50-67-9 and molecular formula C10H12N2O.
  • Used in receptor pharmacology and biochemical assays.
  • Suitable as an analytical standard and for method development.

Catalog No. 50-002-98897 Supplier Medchemexpress LLC Supplier No. HYB1473A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Serotonin (5-hydroxytryptamine) is an endogenous monoamine neurotransmitter and 5-HT receptor agonist commonly used as a biochemical reagent and analytical standard. It is supplied as a high-purity solid suitable for receptor pharmacology, analytical method development, and biochemical assays.

  • High purity (99.26%) suitable for analytical work.
  • Available as a solid in 10 mg quantities.
  • CAS number 50-67-9 and molecular formula C10H12N2O.
  • Used in receptor pharmacology and biochemical assays.
  • Suitable as an analytical standard and for method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.