Learn More
Medchemexpress LLC Serotonin | 50-67-9 | 99.0% | 10 MM 1 ML
SDP
Catalog No. 5000411157
Encompass_Preferred
Serotonin is a monoamine neurotransmitter found in the CNS, acting as an endogenous 5-HT receptor agonist. It also functions as a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM, inhibiting COMT via non-competitive binding to the site bound by catechol substrates.
  • Monoamine neurotransmitter in the CNS
  • Endogenous 5-HT receptor agonist
  • Catechol O-methyltransferase (COMT) inhibitor
  • Inhibits COMT via non-competitive binding
  • Used in various assays including cell proliferation/viability, ELISA, and cell migration/invasion
  • Involved in clinical trials for neurological and mood disorders
Catalog No. 50-004-11157 Supplier Medchemexpress LLC Supplier No. HYB1473A10MM/1ML
Serotonin is a monoamine neurotransmitter found in the CNS, acting as an endogenous 5-HT receptor agonist. It also functions as a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM, inhibiting COMT via non-competitive binding to the site bound by catechol substrates.
  • Monoamine neurotransmitter in the CNS
  • Endogenous 5-HT receptor agonist
  • Catechol O-methyltransferase (COMT) inhibitor
  • Inhibits COMT via non-competitive binding
  • Used in various assays including cell proliferation/viability, ELISA, and cell migration/invasion
  • Involved in clinical trials for neurological and mood disorders