Learn More
Medchemexpress LLC Serotonin | 50-67-9 | 99.0% | 100 MG
SDP
Catalog No. 5000411162
Encompass_Preferred
Serotonin is a monoamine neurotransmitter in the central nervous system (CNS) and acts as an endogenous agonist for 5-HT receptors. It also functions as a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM. This product is for research use only and has not been fully validated for medical applications.
  • Functions as a monoamine neurotransmitter.
  • Acts as an endogenous 5-HT receptor agonist.
  • Inhibits catechol O-methyltransferase (COMT).
  • Suitable for research use only.
  • Not validated for medical applications.
Catalog No. 50-004-11162 Supplier Medchemexpress LLC Supplier No. HYB1473A100MG
Serotonin is a monoamine neurotransmitter in the central nervous system (CNS) and acts as an endogenous agonist for 5-HT receptors. It also functions as a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM. This product is for research use only and has not been fully validated for medical applications.
  • Functions as a monoamine neurotransmitter.
  • Acts as an endogenous 5-HT receptor agonist.
  • Inhibits catechol O-methyltransferase (COMT).
  • Suitable for research use only.
  • Not validated for medical applications.