Learn More
Medchemexpress LLC Serotonin-d4 hydrochloride | 2469263-61-2 | 99.5% | 1 MG
SDP
Catalog No. 5000411417
Encompass_Preferred
Serotonin-d4 hydrochloride is a deuterium-labeled form of Serotonin (5-Hydroxytryptamine) hydrochloride, functioning as a monoamine neurotransmitter in the CNS and an endogenous 5-HT receptor agonist. It also acts as a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM. Stable heavy isotopes, such as deuterium, are incorporated into molecules as tracers for quantitation during development processes, and can influence pharmacokinetic and metabolic profiles. This product is for research use only.
  • Used as a tracer in various applications.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles.
Catalog No. 50-004-11417 Supplier Medchemexpress LLC Supplier No. HYB1473S1MG
Serotonin-d4 hydrochloride is a deuterium-labeled form of Serotonin (5-Hydroxytryptamine) hydrochloride, functioning as a monoamine neurotransmitter in the CNS and an endogenous 5-HT receptor agonist. It also acts as a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM. Stable heavy isotopes, such as deuterium, are incorporated into molecules as tracers for quantitation during development processes, and can influence pharmacokinetic and metabolic profiles. This product is for research use only.
  • Used as a tracer in various applications.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles.