Learn More
Cayman Chemical Serotonin-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 2469263-61-2, 10 mg
SDP
Catalog No. 502992641
Encompass_Preferred
An internal standard for the quantification of serotonin by GC- or LC-MS
This item is not returnable. View return policy
Catalog No. 50-299-2641 Supplier Cayman Chemical Supplier No. 2941510 MG
An internal standard for the quantification of serotonin by GC- or LC-MS