Learn More
Cayman Chemical Serotonin-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 2469263-61-2, 1 mg
SDP
Catalog No. 502990565
Encompass_Preferred
An internal standard for the quantification of serotonin by GC- or LC-MS
Catalog No. 50-299-0565 Supplier Cayman Chemical Supplier No. 294151 MG
An internal standard for the quantification of serotonin by GC- or LC-MS