Learn More
Cayman Chemical SerotonIn-d4hydrochlorIde 5mg
SDP
Catalog No. 502988812
Encompass_Preferred
An internal standard for the quantification of serotonin by GC- or LC-MS
Catalog No. 50-298-8812 Supplier Cayman Chemical Supplier No. 294155 MG
Add to Cart
Add to Cart
An internal standard for the quantification of serotonin by GC- or LC-MS