Learn More
Enzo Life Sciences Serotonin ELISA kit (1x96wells)
SDP
Catalog No. 50950043
Encompass_Preferred
Easy-to-use, well-validated serotonin ELISA kit for hormone research. The Serotonin EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Alternative Name: 5-HT. 5-hydroxytryptamine. Sensitivity: 0.293 ng/ml (range 0.49 - 500 ng/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-950-043 Supplier Enzo Life Sciences Supplier No. ADI900175
Easy-to-use, well-validated serotonin ELISA kit for hormone research. The Serotonin EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Alternative Name: 5-HT. 5-hydroxytryptamine. Sensitivity: 0.293 ng/ml (range 0.49 - 500 ng/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution