Learn More
Abcam Serotonin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1330531X96TESTS
Encompass_Preferred
The Serotonin (5-HT) ELISA kit (colorimetric) is a competitive ELISA designed to quantify serotonin (5-hydroxytryptamine, 5-HT) in serum, plasma, urine, and platelets with a sensitivity of 0.293 ng/mL. - Wide dynamic range – quantifies 0.49–500 ng/mL - No acylation required for standards and sample - Assay platform – microplate for easy setup and reliable results - Cited in over 30 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0928962