Learn More
Apexbio Technology LLC Serotonin HCl(Synonyms: Serotonin hydrochloride, 5-Hydroxytryptamine hydrochloride, 5-HT hydrochloride), 50mg, CAS: 153-98-0.
SDP
Catalog No. 5000060152
Encompass_Preferred
Serotonin hydrochloride (CAS 153-98-0) is a monoamine neurotransmitter naturally derived from tryptophan that functions as an endogenous agonist of serotonin (5-HT) receptors Additionally serotonin hydrochloride acts as a non-competitive inhibitor of the enzyme catechol O-methyltransferase (COMT) exhibiting a binding affinity (Ki) of approximately 44 M Serotonin modulates diverse physiological processes including mood regulation sleep appetite sexual function and gastrointestinal activity Due to its role in mediating neurological and inflammatory pathways serotonin hydrochloride is widely utilized in research studies investigating depression anxiety disorders migraine pathophysiology and inflammation-associated hypersensitivity
Catalog No. 50-000-60152 Supplier Apexbio Technology LLC Supplier No. B183050
Serotonin hydrochloride (CAS 153-98-0) is a monoamine neurotransmitter naturally derived from tryptophan that functions as an endogenous agonist of serotonin (5-HT) receptors Additionally serotonin hydrochloride acts as a non-competitive inhibitor of the enzyme catechol O-methyltransferase (COMT) exhibiting a binding affinity (Ki) of approximately 44 M Serotonin modulates diverse physiological processes including mood regulation sleep appetite sexual function and gastrointestinal activity Due to its role in mediating neurological and inflammatory pathways serotonin hydrochloride is widely utilized in research studies investigating depression anxiety disorders migraine pathophysiology and inflammation-associated hypersensitivity